Learn More
Medchemexpress LLC Rosmarinic acid | 20283-92-5 | 360.31 | 25 MG
SDP
Catalog No. 5000389563
Encompass_Preferred
Rosmarinic acid is an analytical standard intended for research and analytical applications. It is a widespread phenolic ester compound found in plants that inhibits MAO-A, MAO-B, and COMT enzymes.
  • Used for qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits MAO-A, MAO-B, and COMT enzymes
Catalog No. 50-003-89563 Supplier Medchemexpress LLC Supplier No. 25MGHYN0529R
Rosmarinic acid is an analytical standard intended for research and analytical applications. It is a widespread phenolic ester compound found in plants that inhibits MAO-A, MAO-B, and COMT enzymes.
  • Used for qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Inhibits MAO-A, MAO-B, and COMT enzymes