Learn More
Medchemexpress LLC Rosmarinic acid | 20283-92-5 | MFCD00017740 | 99.3% | 360.31 | C18H16O8 | 10 MG
SDP
Catalog No. 5000296885
Encompass_Preferred
Rosmarinic acid is a polyphenolic ester used as an analytical reference material for method development, assay calibration, and compound identification in chromatographic and spectroscopic analyses. It is supplied in a small reference pack suitable for laboratory quality control and analytical workflows.
  • High purity suitable for analytical use.
  • Provided as a small reference quantity for calibration and validation.
  • Soluble in common organic solvents for solution standards.
  • Used for HPLC, LC-MS method development, and identity confirmation.
Catalog No. 50-002-96885 Supplier Medchemexpress LLC Supplier No. HYN0529R10MG
Rosmarinic acid is a polyphenolic ester used as an analytical reference material for method development, assay calibration, and compound identification in chromatographic and spectroscopic analyses. It is supplied in a small reference pack suitable for laboratory quality control and analytical workflows.
  • High purity suitable for analytical use.
  • Provided as a small reference quantity for calibration and validation.
  • Soluble in common organic solvents for solution standards.
  • Used for HPLC, LC-MS method development, and identity confirmation.