Learn More
Selleck Chemical LLC Rosmarinic acid S3612-1g
SDP
Catalog No. 503109279
Encompass_Preferred
Rosmarinic acid (RA Rosemary acid) is a naturally occurring hydroxylated compound It has the ability to block complement fixation inhibit lipoxygenase and cyclooxygenase activity and inhibit the expression of CCL11 and CCR3 by suppressing the IKK- activity in NF- B activation signaling
Catalog No. 50-310-9279 Supplier Selleck Chemical LLC Supplier No. S36121G
Add to Cart
Add to Cart
Rosmarinic acid (RA Rosemary acid) is a naturally occurring hydroxylated compound It has the ability to block complement fixation inhibit lipoxygenase and cyclooxygenase activity and inhibit the expression of CCL11 and CCR3 by suppressing the IKK- activity in NF- B activation signaling