Learn More
Selleck Chemical LLC Rosmarinic acid 25mg 20283-92-5 Rosemary acid
SDP
Catalog No. 501948402
Encompass_Preferred
Rosmarinic acid (RA, Rosemary acid) is a naturally occurring hydroxylated compound. It has the ability to block complement fixation, inhibit lipoxygenase and cyclooxygenase activity and inhibit the expression of CCL11 and CCR3 by suppressing the IKK-? activity in NF-?B activation signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8402 Supplier Selleck Chemical LLC Supplier No. S361225MG
Rosmarinic acid (RA, Rosemary acid) is a naturally occurring hydroxylated compound. It has the ability to block complement fixation, inhibit lipoxygenase and cyclooxygenase activity and inhibit the expression of CCL11 and CCR3 by suppressing the IKK-? activity in NF-?B activation signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.