Learn More
Medchemexpress LLC HY-N0529 100mg , Rosmarinic acid Labiatenic acid CAS:20283-92-5 Purity:98%
SDP
Catalog No. 501493512
Encompass_Preferred
Medchemexpress, HY-N0529 100mg Rosmarinic acid Labiatenic acid CAS:20283-92-5 Formula:C18H16O8 IC50: 50.1 μM (MAO-A), 184.6 μM (MAO-B), 26.7 μM (COMT) Purity:98% Rosmarinic acid (RA) is a widespread phenolic ester compound in the plants. Rosmarinic acid inhibits MAO-A , MAO-B and COMT enzymes with IC 50 s of 50.1, 184.6 and 26.7 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3512 Supplier Medchemexpress LLC Supplier No. HYN0529100MG
Medchemexpress, HY-N0529 100mg Rosmarinic acid Labiatenic acid CAS:20283-92-5 Formula:C18H16O8 IC50: 50.1 μM (MAO-A), 184.6 μM (MAO-B), 26.7 μM (COMT) Purity:98% Rosmarinic acid (RA) is a widespread phenolic ester compound in the plants. Rosmarinic acid inhibits MAO-A , MAO-B and COMT enzymes with IC 50 s of 50.1, 184.6 and 26.7 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.