Learn More
Medchemexpress LLC Rosmarinic acid (standard) | 20283-92-5 | 360.31 | 100 MG
SDP
Catalog No. 5000390097
Encompass_Preferred
Rosmarinic acid (Standard) is an analytical standard intended for research and analytical applications. This widespread phenolic ester compound inhibits MAO-A, MAO-B, and COMT enzymes with IC50s of 50.1, 184.6, and 26.7 μM, respectively. It is for research use only.
  • Analytical standard for Rosmarinic acid
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques
  • Inhibits MAO-A, MAO-B, and COMT enzymes
Catalog No. 50-003-90097 Supplier Medchemexpress LLC Supplier No. HYN0529R100MG
Rosmarinic acid (Standard) is an analytical standard intended for research and analytical applications. This widespread phenolic ester compound inhibits MAO-A, MAO-B, and COMT enzymes with IC50s of 50.1, 184.6, and 26.7 μM, respectively. It is for research use only.
  • Analytical standard for Rosmarinic acid
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques
  • Inhibits MAO-A, MAO-B, and COMT enzymes