Learn More
Medchemexpress LLC Rosmarinic acid (standard) | 20283-92-5 | 360.31 | 50 MG
SDP
Catalog No. 5000389024
Encompass_Preferred
Rosmarinic acid (Standard) is an analytical standard of Rosmarinic acid, a widespread phenolic ester compound found in plants. It is intended for research and analytical applications, and is commonly used in various research experiments such as HPLC, GC, and MS.
  • Inhibits MAO-A, MAO-B, and COMT enzymes with IC50s of 50.1, 184.6, and 26.7 μM.
  • Used as a reference standard for assay purposes.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Commonly used in HPLC, GC, and MS.
Catalog No. 50-003-89024 Supplier Medchemexpress LLC Supplier No. HYN0529R50MG
Rosmarinic acid (Standard) is an analytical standard of Rosmarinic acid, a widespread phenolic ester compound found in plants. It is intended for research and analytical applications, and is commonly used in various research experiments such as HPLC, GC, and MS.
  • Inhibits MAO-A, MAO-B, and COMT enzymes with IC50s of 50.1, 184.6, and 26.7 μM.
  • Used as a reference standard for assay purposes.
  • Suitable for qualitative, quantitative, and methodological research experiments.
  • Commonly used in HPLC, GC, and MS.