Learn More
Medchemexpress LLC HY-A0208 5mg Medchemexpress, Rosoxacin CAS:40034-42-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA02085MG
Encompass_Preferred
Medchemexpress, HY-A0208 5mg Rosoxacin CAS:40034-42-2 Rosoxacin (Acrosoxacin) is a potent and orally active quinolone antibiotic. Rosoxacin (Acrosoxacin) has antibacterial activities against a broad spectrum of Gram negative bacteria including Neisseria gonorrhoeae (MIC90=0.03mg/ml). Rosoxacin has the potential for urinary tract infections treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0090