Learn More
Electron Microscopy Sciences EMS Tissue, 4-1/4 x 5"
SDP
Catalog No. 50949218
Encompass_Preferred
Lens tissue does not lint or scratch and it is free from mineral or vegetable filler.
Size: 4¼ x 5"
Catalog No. 50-949-218 Supplier Electron Microscopy Sciences Supplier No. 71700
Lens tissue does not lint or scratch and it is free from mineral or vegetable filler.
Size: 4¼ x 5"