Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rostafuroxin | 156722-18-8 | 99.0% | 5 MG
SDP

Catalog No. 5000230161
Encompass_Preferred

Rostafuroxin is a digitoxigenin-derived small molecule antagonist of the Na+/K+-ATPase used in preclinical research to study ion-pump signaling and respiratory syncytial virus (RSV)-triggered EGFR phosphorylation.

  • High purity: 99.0%.
  • Molecular formula C23H34O4.
  • Molecular weight 374.5 g/mol.
  • Supplied as 5 MG vial for laboratory use.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-30161 Supplier Medchemexpress LLC Supplier No. HY122835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rostafuroxin is a digitoxigenin-derived small molecule antagonist of the Na+/K+-ATPase used in preclinical research to study ion-pump signaling and respiratory syncytial virus (RSV)-triggered EGFR phosphorylation.

  • High purity: 99.0%.
  • Molecular formula C23H34O4.
  • Molecular weight 374.5 g/mol.
  • Supplied as 5 MG vial for laboratory use.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.