Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rostafuroxin | 156722-18-8 | 99.0% | 50 MG
SDP

Catalog No. 5000231981
Encompass_Preferred

Rostafuroxin is a small-molecule ATP1A1 (Na+/K+-ATPase) antagonist supplied as a research-grade analytical standard for laboratory use. It is used to study Na+/K+-ATPase modulation, hypertension mechanisms, and related signaling pathways.

  • High purity suitable for analytical and research applications.
  • CAS 156722-18-8 and well-defined molecular identity.
  • Molecular formula C23H34O4 and molecular weight 374.5 g/mol.
  • Soluble in DMSO; typical handling for small-molecule standards.
  • Pack size commonly supplied as 50 MG.

Catalog No. 50-002-31981 Supplier Medchemexpress LLC Supplier No. HY1228350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rostafuroxin is a small-molecule ATP1A1 (Na+/K+-ATPase) antagonist supplied as a research-grade analytical standard for laboratory use. It is used to study Na+/K+-ATPase modulation, hypertension mechanisms, and related signaling pathways.

  • High purity suitable for analytical and research applications.
  • CAS 156722-18-8 and well-defined molecular identity.
  • Molecular formula C23H34O4 and molecular weight 374.5 g/mol.
  • Soluble in DMSO; typical handling for small-molecule standards.
  • Pack size commonly supplied as 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.