Learn More
Apexbio Technology LLC Rosuvastatin(Synonyms: Crestor, Rosuvastatin calcium, ZD4522, Rosulip, Rovista, Rosuvas, R2, Rosucard, Rosudapin, Roswera), 100mg, CAS: 287714-41-4.
SDP
Catalog No. 5000060404
Encompass_Preferred
Rosuvastatin (CAS 287714-41-4) is a small molecule that acts as a competitive inhibitor of the enzyme hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase a critical enzyme catalyzing the rate-limiting step in cholesterol biosynthesis The compound exhibits potent inhibitory activity demonstrated by an IC50 value of approximately 11 nM By inhibiting HMG-CoA reductase rosuvastatin effectively reduces the biosynthesis of cholesterol facilitating its application in research focused on lipid metabolism cardiovascular diseases and related cellular pathways
Catalog No. 50-000-60404 Supplier Apexbio Technology LLC Supplier No. B1123100
Rosuvastatin (CAS 287714-41-4) is a small molecule that acts as a competitive inhibitor of the enzyme hydroxymethylglutaryl-coenzyme A (HMG-CoA) reductase a critical enzyme catalyzing the rate-limiting step in cholesterol biosynthesis The compound exhibits potent inhibitory activity demonstrated by an IC50 value of approximately 11 nM By inhibiting HMG-CoA reductase rosuvastatin effectively reduces the biosynthesis of cholesterol facilitating its application in research focused on lipid metabolism cardiovascular diseases and related cellular pathways