Learn More
TARGETMOL CHEMICALS INC ROSUVASTATIN 25MG
SDP
Catalog No. 502577327
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Rosuvastatin (ZD 4522) is an antilipemic agent that competitively inhibits hydroxymethyl-glutaryl-coenzyme A (HMG-CoA) reductase. HMG-CoA reductase catalyzes the conversion of HMG-CoA to mevalonic acid the rate-limiting step in cholesterol biosynthesis. Rosuvastatin belongs to a class of medications called statins and is used to reduce plasma cholesterol levels and prevent cardiovascular disease. purity: 99%
Catalog No. 50-257-7327 Supplier TARGETMOL CHEMICALS INC Supplier No. T167625MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Rosuvastatin (ZD 4522) is an antilipemic agent that competitively inhibits hydroxymethyl-glutaryl-coenzyme A (HMG-CoA) reductase. HMG-CoA reductase catalyzes the conversion of HMG-CoA to mevalonic acid the rate-limiting step in cholesterol biosynthesis. Rosuvastatin belongs to a class of medications called statins and is used to reduce plasma cholesterol levels and prevent cardiovascular disease. purity: 99%