Learn More
Medchemexpress LLC Rosuvastatin calcium | 147098-20-2 | MFCD04112702 | 99.8% | 500.57 g/mol | C22H27Ca0.5FN3O6S | 10 MG
SDP
Catalog No. 5000298711
Encompass_Preferred
Rosuvastatin calcium is the hemicalcium salt of rosuvastatin, a potent competitive HMG-CoA reductase inhibitor used in research to study cholesterol metabolism, lipid-lowering effects, and cardiac electrophysiology. Supplied as a solid and as DMSO solutions, it is used for in vitro assays exploring statin pharmacology and hERG-related effects.
  • Competitive HMG-CoA reductase inhibitor (IC50 ≈ 11 nM).
  • Potent blocker of hERG current (IC50 ≈ 195 nM).
  • High purity: 99.77% (supplier listed).
  • CAS number 147098-20-2.
  • Supplied as powder and 10 mM in DMSO solution for in vitro use.
  • Commonly used to study LDL and triglyceride reduction and electrophysiological effects.
Catalog No. 50-002-98711 Supplier Medchemexpress LLC Supplier No. HY1750410MG
Rosuvastatin calcium is the hemicalcium salt of rosuvastatin, a potent competitive HMG-CoA reductase inhibitor used in research to study cholesterol metabolism, lipid-lowering effects, and cardiac electrophysiology. Supplied as a solid and as DMSO solutions, it is used for in vitro assays exploring statin pharmacology and hERG-related effects.
  • Competitive HMG-CoA reductase inhibitor (IC50 ≈ 11 nM).
  • Potent blocker of hERG current (IC50 ≈ 195 nM).
  • High purity: 99.77% (supplier listed).
  • CAS number 147098-20-2.
  • Supplied as powder and 10 mM in DMSO solution for in vitro use.
  • Commonly used to study LDL and triglyceride reduction and electrophysiological effects.