Learn More
Selleck Chemical LLC Rosuvastatin Calcium 50mg 147098-20-2 ZD4522
SDP
Supplier:  Selleck Chemical LLC S216950MG
Encompass_Preferred
Rosuvastatin Calcium (ZD4522) is a competitive inhibitor of HMG-CoA reductase with IC50 of 11 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-665-6