Learn More
Medchemexpress LLC Rosuvastatin-d3 sodium | 1279031-70-7 | 99.1% | 506.54 | 1 MG
SDP
Catalog No. 5000387009
Encompass_Preferred
Rosuvastatin-d3 sodium is a deuterium labeled Rosuvastatin. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. This product is for research use only and not for sale to patients.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for therapeutic drug monitoring
Catalog No. 50-003-87009 Supplier Medchemexpress LLC Supplier No. HY17504BS1MG
Rosuvastatin-d3 sodium is a deuterium labeled Rosuvastatin. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. This product is for research use only and not for sale to patients.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for therapeutic drug monitoring