Learn More
Medchemexpress LLC Rosuvastatin-d3 sodium | 1279031-70-7 | 99.1% | 506.54 | 5 MG
SDP
Catalog No. 5000386781
Encompass_Preferred
Rosuvastatin-d3 (sodium) is deuterium labeled Rosuvastatin. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86781 Supplier Medchemexpress LLC Supplier No. HY17504BS5MG
Add to Cart
Add to Cart
Rosuvastatin-d3 (sodium) is deuterium labeled Rosuvastatin. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.