Learn More
Cayman Chemical Rosuvastatin-d6 (sodium salt), Purity: ≥98%, CAS: 2070009-41-3, 1 mg
SDP
Catalog No. 502987991
Encompass_Preferred
Rosuvastatin-d6 is intended for use as an internal standard for the quantification of rosuvastatin by GC- or LC-MS.
This item is not returnable. View return policy
Catalog No. 50-298-7991 Supplier Cayman Chemical Supplier No. 233591 MG
Rosuvastatin-d6 is intended for use as an internal standard for the quantification of rosuvastatin by GC- or LC-MS.