Learn More
Cayman Chemical RosuvastatIncalcIum salt 10mg
SDP
Catalog No. 502967909
Encompass_Preferred
An HMG-CoA reductase inhibitor (IC50 = 5 nM); inhibits cholesterol synthesis in isolated rat hepatocytes (IC50 = 0.16 nM); reduces plasma total cholesterol, triglyceride, LDL-C, and oxidized LDL-C levels in Ldlr-/- mice fed a high-fat diet at 10 mg/kg; decreases the area of aortic atherosclerotic lesions in the same model
Catalog No. 50-296-7909 Supplier Cayman Chemical Supplier No. 1881310 MG
An HMG-CoA reductase inhibitor (IC50 = 5 nM); inhibits cholesterol synthesis in isolated rat hepatocytes (IC50 = 0.16 nM); reduces plasma total cholesterol, triglyceride, LDL-C, and oxidized LDL-C levels in Ldlr-/- mice fed a high-fat diet at 10 mg/kg; decreases the area of aortic atherosclerotic lesions in the same model