Learn More
AVANTOR MASTERFLEX (VWR LLC) Rotary Drum Pump, PP/PTFE, 8 GPM; 2"" IPS Bung
Catalog No. NC1407000
Encompass
Rotary Drum Pump, PP/PTFE, 8 GPM; 2" IPS Bung, Dispense a variety of liquids compatible with polypropylene, Ryton PPS, cast iron, aluminum, stainless steel, or PTFELightweight and easy to useSmooth, non-pulsating flowOperate either in forward or reverse direction with a discharge hose
Catalog No. NC1407000 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0651000
Rotary Drum Pump, PP/PTFE, 8 GPM; 2" IPS Bung, Dispense a variety of liquids compatible with polypropylene, Ryton PPS, cast iron, aluminum, stainless steel, or PTFELightweight and easy to useSmooth, non-pulsating flowOperate either in forward or reverse direction with a discharge hose