Learn More
Alkali Scientific Roto-Mini Plus, Variable Speed Rotator with tube holders
SDP
Catalog No. 502335796
Encompass_Preferred
Roto-Mini series of rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including re-suspension, protein binding, nucleic acid purification, etc. the mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. The Roto-Mini rotator is also available in a “Plus” configuration, providing the following additional features: Variable speed: 5 to 70rpm “Mix” function: Rotating in combination w/ oscillations (increased mixing) “Pause” function: Rotating with intermittent “stops” during rotation “Rock” function: Back and forth, gentle rocking motion (see-saw) The Plus models also offer an additional rocking platform (4.5 x 3 in.), as well as a 96 x 0.2ml tube/strip/plate holder for mixing PCR vessels in the “Mix” (oscillating) mode. Specifications: Speed : 24rpm Spe
Catalog No. 50-233-5796 Supplier Alkali Scientific Supplier No. RS3024
Roto-Mini series of rotating mixers provide a wide range of mixing capabilities. Suitable for an array of laboratory applications including re-suspension, protein binding, nucleic acid purification, etc. the mixers include interchangeable tube holders for accommodating all common tube sizes from 0.5ml to 50ml. The motion can be adjusted from vertical (end over end) mixing, to horizontal (rolling or hybridization style) mixing. The Roto-Mini rotator is also available in a “Plus” configuration, providing the following additional features: Variable speed: 5 to 70rpm “Mix” function: Rotating in combination w/ oscillations (increased mixing) “Pause” function: Rotating with intermittent “stops” during rotation “Rock” function: Back and forth, gentle rocking motion (see-saw) The Plus models also offer an additional rocking platform (4.5 x 3 in.), as well as a 96 x 0.2ml tube/strip/plate holder for mixing PCR vessels in the “Mix” (oscillating) mode. Specifications: Speed : 24rpm Spe