Learn More
Organomation 8 Position ROT-X-TRACT-S Soxhlet Extractor
SDP
Catalog No. NC2236349
Encompass_Preferred
Soxhlet extraction is ideal for extracting soluble analytes from a solid sample into an organic solvent. Unlike traditional wire or sand baths which are used to heat receiving flasks, the ROT-X-TRACT-S uses a water steam bath which provides gentle even heat to delicate samples.The ROT-X-TRACT-S rotary Soxhlet extractors can accommodate up to 8 or 10 Soxhlet extractors. The bath cover disc holes can accommodate round sample flasks from 125 ml-1 L. Models 13070 and 13090 can accommodate smaller 250 mL flasks, while model 13008 can accommodate larger 500 mL flasks. A wide range of sample sizes and quantities make this instrument ideal for Soxhlet extraction of samples in any lab.
This item is not returnable. View return policy
Catalog No. NC2236349 Supplier Organomation Supplier No. 13008
Add to Cart
Add to Cart
Request a Quote
Soxhlet extraction is ideal for extracting soluble analytes from a solid sample into an organic solvent. Unlike traditional wire or sand baths which are used to heat receiving flasks, the ROT-X-TRACT-S uses a water steam bath which provides gentle even heat to delicate samples.The ROT-X-TRACT-S rotary Soxhlet extractors can accommodate up to 8 or 10 Soxhlet extractors. The bath cover disc holes can accommodate round sample flasks from 125 ml-1 L. Models 13070 and 13090 can accommodate smaller 250 mL flasks, while model 13008 can accommodate larger 500 mL flasks. A wide range of sample sizes and quantities make this instrument ideal for Soxhlet extraction of samples in any lab.