Learn More
Electron Microscopy Sciences Rota-Rack for Large Tubes 1 each
SDP
Catalog No. 5034155
Encompass_Preferred
Connect up to 4 interlocking racks and rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry! Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable. Holds nine 50 ml tubes, eight 15 ml tubes, six 20 mm tubes, or ten 10-13 mm tubes.
Catalog No. 50-341-55 Supplier Electron Microscopy Sciences Supplier No. 6160410
Add to Cart
Add to Cart
Connect up to 4 interlocking racks and rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry! Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable. Holds nine 50 ml tubes, eight 15 ml tubes, six 20 mm tubes, or ten 10-13 mm tubes.