Learn More
Electron Microscopy Sciences Rota-Rack for Microtubes
SDP
Catalog No. 5034156
Encompass_Preferred
Connect up to 4 interlocking racks and rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry! Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable. Holds 6 15 ml, 9 1.5/2.0 ml, 12 0.5/0.6 ml , or 32 0.2 ml PCR* tubes or 4 8-tube strips.
Catalog No. 50-341-56 Supplier Electron Microscopy Sciences Supplier No. 6160420
Connect up to 4 interlocking racks and rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry! Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable. Holds 6 15 ml, 9 1.5/2.0 ml, 12 0.5/0.6 ml , or 32 0.2 ml PCR* tubes or 4 8-tube strips.