Description
Widely used in neuroscience, pharmacology, toxicology, and disease-model research, the system enables researchers to evaluate the effects of drugs, neurological disorders, brain injury, aging, and other interventions on motor performance. The latest generation of Rota Rod incorporates advanced speed protocols, rocking modes, and touchscreen operation to deliver greater experimental flexibility and sensitivity. The Panlab Rota Rod is trusted by behavioral neuroscience laboratories, phenotyping facilities, and preclinical research centers for robust motor function assessment.
- Comprehensive assessment of motor performance, ideal for studies involving neurodegenerative diseases, motor phenotyping, drug screening, aging research, and neurological injury models.
- Extended speed range for enhanced experimental sensitivity enabling detection of subtle motor impairments and performance differences that may be missed using conventional protocols.
- Constant speed and acceleration (ramp) testing supporting standardized and reproducible experiments.
- Advanced speed protocol and rocking modes: allowing researchers to challenge animals under a broader range of conditions and better characterize motor deficits.
- Automatic measurement of key performance parameters proving objective, quantitative data for evaluating motor function and treatment effects.
- Intuitive touchscreen user interface Touchscreen display providing clear visualization of speed and timing for each lane and simplifying mode selection, speed adjustment, directly from the main screen.
- Flexible configurations for mice and rats supporting a wide range of study designs and laboratory requirements.
- Accurate and reliable operation delivering reproducible results essential for preclinical behavioral research.
- Automatic data storage and software integration enabling convenient data transfer, storage, and analysis.
- Research-proven platform cited in thousands of published scientific studies worldwide.
- Applications: Motor coordination and balance studies, Endurance and fatigue resistance testing, Neurodegenerative disease models (e.g., Parkinson's and Huntington's disease), Drug efficacy and safety screening, Aging research, Neurological injury and recovery studies, Behavioral phenotyping in mice and rats
Specifications
Specifications
| Product Type | Rota Rod |
| Animal | Rat, Mice |
| Number of Lanes | 4 |
| Display Type | Touchscreen |
| Includes | Rota Rod with touchscreen controller, rod/s, cables and connectors, Sedacom software |