Learn More
Electron Microscopy Sciences Rota Filler 1000™ Replacement Silicone Pipette Adapter
SDP
Catalog No. 501929502
Encompass_Preferred
The Rota Filler 1000 is a manual pipette controller uniquely designed for one-handed operation. To operate, simply squeeze the main bulb, then push the toggle forward to aspirate, or back to dispense. Pushing the blowout bulb will expel any liquid remaining in the pipette.
It features a replaceable tapered silicone pipette adapter to allow a large range of pipette sizes and makes to be used. Using common and readily available replacement filters will help reduce total cost of ownership over time. Includes a magnetic/wall mountable holder and 0.45 um filter. For use with glass and plastic pipettes 1 t 50 ml
Catalog No. 50-192-9502 Supplier Electron Microscopy Sciences Supplier No. 7097503
The Rota Filler 1000 is a manual pipette controller uniquely designed for one-handed operation. To operate, simply squeeze the main bulb, then push the toggle forward to aspirate, or back to dispense. Pushing the blowout bulb will expel any liquid remaining in the pipette.
It features a replaceable tapered silicone pipette adapter to allow a large range of pipette sizes and makes to be used. Using common and readily available replacement filters will help reduce total cost of ownership over time. Includes a magnetic/wall mountable holder and 0.45 um filter. For use with glass and plastic pipettes 1 t 50 ml