Learn More
Hettich Centrifuges ROTANTA460,(196)13(48)16,BioPk
SDP
Supplier:  Hettich Centrifuges 460BLOODHCBC
Encompass_Preferred
Hettich ROTANTA 460 BLOOD TUBE PACKAGE HIGH CAPACITY BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 13 x 75/100 mm, (4) Adapters 16 x 75/100, (4) Bio-containment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7579
Add to Cart
Request a Quote