Learn More
Hettich Centrifuges ROTANTA460R,(108)13(88)16,BCPk
SDP
Catalog No. 502537567 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTANTA 460 R BLOOD TUBE PACKAGE 4B BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 13 x 75/100 mm, (4) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7567 Supplier Hettich Centrifuges Supplier No. 460RBLOOD4BBC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTANTA 460 R BLOOD TUBE PACKAGE 4B BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 13 x 75/100 mm, (4) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years