Learn More
Hettich Centrifuges ROTANTA460R,(28)15(14)50,BCPackage
SDP
Catalog No. 502537560 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTANTA 460 R CELL CULTURE PACKAGE 2 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7560 Supplier Hettich Centrifuges Supplier No. 460RCELLCULTURE2BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTANTA 460 R CELL CULTURE PACKAGE 2 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (2) Adapters 15 mL, (2) Adapters 50 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years