Learn More
Hettich Centrifuges ROTANTA460R,(42)13(38)16,BioPk
SDP
Catalog No. 502537569 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTANTA 460 R BLOOD TUBE PACKAGE 2 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (2) Adapters 13 x 75/100 mm, (2) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7569 Supplier Hettich Centrifuges Supplier No. 460RBLOOD2BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTANTA 460 R BLOOD TUBE PACKAGE 2 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (2) Adapters 13 x 75/100 mm, (2) Adapters 16 x 75/100, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years