Learn More
Hettich Centrifuges ROTANTA460R,(4)250 mL,Bio,Package
SDP
Catalog No. 502537562 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTANTA 460 R BOTTLE PACKAGE 250 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 250/290 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7562 Supplier Hettich Centrifuges Supplier No. 460RBOTTLE250BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTANTA 460 R BOTTLE PACKAGE 250 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 250/290 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years