Learn More
Hettich Centrifuges ROTANTA460R,(96)15(40)50,BCPackage
SDP
Supplier:  Hettich Centrifuges 460RCELLCULTUREHCBC
Encompass_Preferred
Hettich ROTANTA 460 R CELL CULTURE PACKAGE HIGH CAPACITY BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 15 mL, (4) Adapters 50 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7557
Add to Cart
Request a Quote