Learn More
Hettich Centrifuges ROTANTA460RF(56)15(28)50,BCPackage
SDP
Catalog No. 502537546 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich ROTANTA 460 RF CELL CULTURE PACKAGE 4 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 15 mL, (4) Adapters 50 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7546 Supplier Hettich Centrifuges Supplier No. 460RFCELLCULTURE4BC
Add to Cart
Add to Cart
Request a Quote
Hettich ROTANTA 460 RF CELL CULTURE PACKAGE 4 BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters 15 mL, (4) Adapters 50 mL, (4) Biocontainment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years