Learn More
Heidolph™ Rotary Button for Laborota 20, Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900937 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-937 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-937 Supplier Heidolph™ Supplier No. 036259100
Add to Cart
Add to Cart
Rotary button allows user to lock/unlock rotary evaporator flask rotation.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator
Specifications
Type Button
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov