Learn More
Chemglass Life Sciences Bump Trap, 100mL, Rotary Evaporator, Modified, 29/42 Top Outer, 29/42 Lower Inner
SDP
Catalog No. 501531073
Encompass_Preferred
Bump Trap, 100mL, Rotary Evaporator, Modified, 29/42 Top Outer, 29/42 Lower Inner. Similar to CG-1320, but with the addition of two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Traps have a 29/42 top outer joint and a 29/42 lower inner joint.
Catalog No. 50-153-1073 Supplier Chemglass Life Sciences Supplier No. CG1321102
Bump Trap, 100mL, Rotary Evaporator, Modified, 29/42 Top Outer, 29/42 Lower Inner. Similar to CG-1320, but with the addition of two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Traps have a 29/42 top outer joint and a 29/42 lower inner joint.