Learn More
Chemglass Life Sciences Bump Trap, 250mL, Rotary Evaporator, 29/42 Top Outer, 29/42 Lower Inner
SDP
Catalog No. 50873439
Encompass_Preferred
Bump Trap, 250mL, Rotary Evaporator, 29/42 Top Outer, 29/42 Lower Inner. Used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of “bumping.” Traps have a 24/40 top outer joint and the listed size of lower inner joint.
Catalog No. 50-873-439 Supplier Chemglass Life Sciences Supplier No. CG1320252
Bump Trap, 250mL, Rotary Evaporator, 29/42 Top Outer, 29/42 Lower Inner. Used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of “bumping.” Traps have a 24/40 top outer joint and the listed size of lower inner joint.