Learn More
Chemglass Life Sciences 24/40 Vapor Tube/Trap Adapter, Fritted
SDP
Catalog No. 501531070
Encompass_Preferred
24/40 Vapor Tube/Trap Adapter, Fritted. Vapor tube/bump trap adapter is designed to be used with the Bchi R-200/205 rotary evaporators. This adapter combines a vapor tube adapter and a bump trap. Trap has an extra coarse fritted disc sealed between the lower inner joint and the top flange. Adapters have a 50mm OD top flange with a tapered interior to fit the Bchi sealing ring. Lower portion of the adapter has the listed standard taper inner joint. Each adapter is supplied complete with the corresponding size Keck clip.
Catalog No. 50-153-1070 Supplier Chemglass Life Sciences Supplier No. CG1317C10
24/40 Vapor Tube/Trap Adapter, Fritted. Vapor tube/bump trap adapter is designed to be used with the Bchi R-200/205 rotary evaporators. This adapter combines a vapor tube adapter and a bump trap. Trap has an extra coarse fritted disc sealed between the lower inner joint and the top flange. Adapters have a 50mm OD top flange with a tapered interior to fit the Bchi sealing ring. Lower portion of the adapter has the listed standard taper inner joint. Each adapter is supplied complete with the corresponding size Keck clip.