Learn More
Chemglass Life Sciences Replacement Steam Duct for Buchi R-200/R-205 Rotary Evaporators, 29/42 Steam Duct
SDP
Catalog No. CG1317A12
Encompass_Preferred
Replacement vapor (steam) tube adapter for Bchi R-200/205 rotary evaporators. Adapters have a 50mm OD top flange with a tapered interior to fit the Bchi sealing ring. Lower portion of the adapter has the listed standard taper inner joint. Each tube is supplied complete with corresponding size Keck clip.NOTE: ITEMS WILL REPLACE THE BUCHI PART NUMBERS AS LISTED. CHEMGLASS ITEMS ARE TO BE USED WITH THE KECK CLIP, NOT THE BUCHI COMBI-CLIP.
Catalog No. CG1317A12 Supplier Chemglass Life Sciences Supplier No. CG1317A12
Replacement vapor (steam) tube adapter for Bchi R-200/205 rotary evaporators. Adapters have a 50mm OD top flange with a tapered interior to fit the Bchi sealing ring. Lower portion of the adapter has the listed standard taper inner joint. Each tube is supplied complete with corresponding size Keck clip.NOTE: ITEMS WILL REPLACE THE BUCHI PART NUMBERS AS LISTED. CHEMGLASS ITEMS ARE TO BE USED WITH THE KECK CLIP, NOT THE BUCHI COMBI-CLIP.