Learn More
Chemglass Life Sciences Rotary Evaporator Complete, 5L, 115V
SDP
Catalog No. 501531078
Encompass_Preferred
Rotary Evaporator Complete, 5L, 115V. Features: Large PTFE coated heating bath With reflux valve for distillation under total reflux Unique design vapor seal maintains vacuum 5L Evaporating Flask Capacity with flat flange With feeding valve and bottom drain valve for continuous operation without interruption of vacuum Motorized lift with push button control. Unique 5L Rotary Evaporator design fills the opening between benchtop and industrial evaporators systems. Features a large bath for a 5L flask. Continuous reflux with a 3L receiving flask and a reflux valve and a bottom drain valve for the receiving flask. Motorized lift with push button control. Heating temperature range from ambient to 180C.
Catalog No. 50-153-1078 Supplier Chemglass Life Sciences Supplier No. CG1334X50
Add to Cart
Add to Cart
Request a Quote
Rotary Evaporator Complete, 5L, 115V. Features: Large PTFE coated heating bath With reflux valve for distillation under total reflux Unique design vapor seal maintains vacuum 5L Evaporating Flask Capacity with flat flange With feeding valve and bottom drain valve for continuous operation without interruption of vacuum Motorized lift with push button control. Unique 5L Rotary Evaporator design fills the opening between benchtop and industrial evaporators systems. Features a large bath for a 5L flask. Continuous reflux with a 3L receiving flask and a reflux valve and a bottom drain valve for the receiving flask. Motorized lift with push button control. Heating temperature range from ambient to 180C.