Learn More
Chemglass Life Sciences Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 29/42 Lower Inner
SDP
Catalog No. CG132324
Encompass_Preferred
Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 29/42 Lower Inner. Trap has an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping. All traps have a 24/40 top outer joint and the listed size of the lower inner joint.
Catalog No. CG132324 Supplier Chemglass Life Sciences Supplier No. CG132324
Bump Trap, Fritted, X-Coarse, Rotary Evaporator, 24/40 Top Outer, 29/42 Lower Inner. Trap has an extra coarse fritted disc sealed between the lower inner and top outer joint to prevent contents of the flask from entering into the condenser in the event of bumping. All traps have a 24/40 top outer joint and the listed size of the lower inner joint.