Learn More
Chemglass Life Sciences Rotary Evaporator, Dry Ice Condenser, 110V, 50/60Hz, US Plug
SDP
Catalog No. 501531076
Encompass_Preferred
Rotary Evaporator, Dry Ice Condenser, 110 Volt. The Chemglass Rotary Evaporator has a large easy-to-read digital LCD screen displaying the temperature, rotation speed and timer. Features a large 5L water bath and has a temperature range from ambient to 180C. Features: Easy to Read Large Digital LCD Display Shows Temperature, Rotation Speed and Timer Large 5L Heating Bath with a Temperature Range of Ambient to 180C Motorized Lift with Safety Automatic Release of the Evaporating Flask to the Top Position in case of Power Failure Speed Range from 20 to 280 RPM Timer Function for Precise Control Processing Chemical-Resistant PTFE Seals and Patented Pressure Springs Provide Excellent Sealing PC Control and Documentation of All Parameters via USB Precise Temperature Control and Adjustable Safety Circuit Quick Release Connectors for Easy Flask Removal
Catalog No. 50-153-1076 Supplier Chemglass Life Sciences Supplier No. CG133420
Add to Cart
Add to Cart
Request a Quote
Rotary Evaporator, Dry Ice Condenser, 110 Volt. The Chemglass Rotary Evaporator has a large easy-to-read digital LCD screen displaying the temperature, rotation speed and timer. Features a large 5L water bath and has a temperature range from ambient to 180C. Features: Easy to Read Large Digital LCD Display Shows Temperature, Rotation Speed and Timer Large 5L Heating Bath with a Temperature Range of Ambient to 180C Motorized Lift with Safety Automatic Release of the Evaporating Flask to the Top Position in case of Power Failure Speed Range from 20 to 280 RPM Timer Function for Precise Control Processing Chemical-Resistant PTFE Seals and Patented Pressure Springs Provide Excellent Sealing PC Control and Documentation of All Parameters via USB Precise Temperature Control and Adjustable Safety Circuit Quick Release Connectors for Easy Flask Removal