Learn More
Chemglass Life Sciences Rotary Evaporator Flask Only, 5 L - Replacement for Chemglass 5 L Rotary Evaporator
SDP
Catalog No. 502733596
Encompass_Preferred
Replacement 5 L rotary evaporator flask (round-bottom) designed for use with compatible 5 L rotary evaporator systems. Provides durable glass construction and a flat flange for secure sealing.
  • Capacity 5 L.
  • Material glass construction.
  • Flange type flat flange for secure sealing.
  • Intended use replacement evaporation flask for 5 L rotary evaporators.
  • Compatible with standard rotary evaporator condensers and receiving flasks.
Catalog No. 50-273-3596 Supplier Chemglass Life Sciences Supplier No. CG1334X52
Add to Cart
Add to Cart
Replacement 5 L rotary evaporator flask (round-bottom) designed for use with compatible 5 L rotary evaporator systems. Provides durable glass construction and a flat flange for secure sealing.
  • Capacity 5 L.
  • Material glass construction.
  • Flange type flat flange for secure sealing.
  • Intended use replacement evaporation flask for 5 L rotary evaporators.
  • Compatible with standard rotary evaporator condensers and receiving flasks.