Learn More
Chemglass Life Sciences Motor Kit, 24/40 Bushing Adapter, 3-Piece Tool Kit (CG-1981-C-24), Motor w/ 2 Lithium Batteries
SDP
Catalog No. 501354679
Encompass_Preferred
Motor Kit, 24/40 Bushing Adapter, 3-Piece Tool Kit (CG-1981-C-24), Motor w/ 2 Batteries. Easily remove dried-on solids from the interior walls of round bottom flasks without scratching or harming your flask!Chem-spin scraper tools provide an easy method of removing the majority of desired, dried-on solids from the interior walls of round bottom flasks. Motor driven, the spinning chain rapidly and effectively brushes the surface as the loose solids collect on the bottom. Minimal physical effort is required. Tedious, time-consuming, manual scraping with a spatula is eliminated.Motor Kits include everything you need to get started: rotary scraper tools, glass bushing adapters, motors and two 12V batteries.
Catalog No. 50-135-4679 Supplier Chemglass Life Sciences Supplier No. CG1981C28
Motor Kit, 24/40 Bushing Adapter, 3-Piece Tool Kit (CG-1981-C-24), Motor w/ 2 Batteries. Easily remove dried-on solids from the interior walls of round bottom flasks without scratching or harming your flask!Chem-spin scraper tools provide an easy method of removing the majority of desired, dried-on solids from the interior walls of round bottom flasks. Motor driven, the spinning chain rapidly and effectively brushes the surface as the loose solids collect on the bottom. Minimal physical effort is required. Tedious, time-consuming, manual scraping with a spatula is eliminated.Motor Kits include everything you need to get started: rotary scraper tools, glass bushing adapters, motors and two 12V batteries.