Learn More
V&P Scientific Inc MultiStirrus™ Tumble Stirrer, Manual Control
Catalog No. NC1314684
Encompass
STIRRER, Vertical, Lateral and Conventional Tumble, DC Motor, 1250 Max RPM, 25.4mm Long Stirring Area, 48 MGO NdFeB Magnetic Drive Cylinder, Connection Cord Outlet on side, 120/220 Volts, 50/60Hz, CE Compliant
Catalog No. NC1314684 Supplier V&P Scientific Inc Supplier No. VP 710D3
STIRRER, Vertical, Lateral and Conventional Tumble, DC Motor, 1250 Max RPM, 25.4mm Long Stirring Area, 48 MGO NdFeB Magnetic Drive Cylinder, Connection Cord Outlet on side, 120/220 Volts, 50/60Hz, CE Compliant