Learn More
RWD LIFE SCIENCE INC Rotary Microtome S700 Semi-Automatic
SDP
Catalog No. NC3404380
Encompass_Preferred
RWD rotary microtome, certified by FDA and ISO13485, is designed for PPFP tissue sections, histopathology, and anatomical pathology. It ensures precise, safe cuts with an ergonomic design for comfortable use. The system delivers reproducible, high-quality sections as thin as 0.5 um. The 2-in-1 blade holder accommodates both high-profile and low-profile blades, and it features patented pointer technology and base scale, which enable precise sample positioning. Its handwheel double-lock system, knife ejector, and guard for enhanced safety. A 5-inch touch screen with history recording simplifies tracking and troubleshooting. The ergonomic side knob and easy-to-clean waste tank ensure convenient operation and maintenance.
Catalog No. NC3404380 Supplier RWD LIFE SCIENCE INC Supplier No. S700
Add to Cart
Add to Cart
Request a Quote
RWD rotary microtome, certified by FDA and ISO13485, is designed for PPFP tissue sections, histopathology, and anatomical pathology. It ensures precise, safe cuts with an ergonomic design for comfortable use. The system delivers reproducible, high-quality sections as thin as 0.5 um. The 2-in-1 blade holder accommodates both high-profile and low-profile blades, and it features patented pointer technology and base scale, which enable precise sample positioning. Its handwheel double-lock system, knife ejector, and guard for enhanced safety. A 5-inch touch screen with history recording simplifies tracking and troubleshooting. The ergonomic side knob and easy-to-clean waste tank ensure convenient operation and maintenance.