Learn More
Cole-Parmer Fluke 9040 Digital Phase Rotation Indicator
Catalog No. 50993273 Shop All Cole-Parmer Products
Encompass
The Fluke 9040 is effective for measuring phase rotation in all areas where three-phase supplies are used to feed motors, drives and electrical systems. This unit is a rotary field indicator and can provide clear indication of the three-phase via an LCD display adn the phase rotation direction to determine correct connections. It allows rapid determination of phase sequence and has a voltage (up to 700V) and frequency range suitable for commercial and industrial applications. The included test probes have a variable clamping range for safe contact, also in industrial sockets.
FEATURES
  • 3 phase indication
  • Indication of phase rotation
  • Clear LCD display
  • No battery required
Catalog No. 50-993-273 Supplier Cole-Parmer Supplier No. 2002141
The Fluke 9040 is effective for measuring phase rotation in all areas where three-phase supplies are used to feed motors, drives and electrical systems. This unit is a rotary field indicator and can provide clear indication of the three-phase via an LCD display adn the phase rotation direction to determine correct connections. It allows rapid determination of phase sequence and has a voltage (up to 700V) and frequency range suitable for commercial and industrial applications. The included test probes have a variable clamping range for safe contact, also in industrial sockets.
FEATURES
  • 3 phase indication
  • Indication of phase rotation
  • Clear LCD display
  • No battery required