Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slice™ Rotary Scissors

Catalog No. 18222845
Change view
Click to view available options
Length (English):
5.51 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
18-222-845 5.51 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-222-845 Supplier Slice™ Supplier No. 10598
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Slice™ Rotary Scissors with bladeless design are ideal for cutting through thin, stiff materials quickly and ergonomically.

  • Blade-free scissors
  • Durable reinforced nylon handle with soft-touch grip lining
  • Safer than traditional scissors
  • Ambidextrous design
  • Reduces injuries, lowers costs
  • Oil, lubricant, and maintenance free
  • Lightweight and sturdy

Specifications

Product Type Rotary
Length (English) 5.51 in.
Length (Metric) 140 mm
Material TPE, Stainless Steel, GCr15
Description Bladeless
For Use With (Application) Cutting stiff thin materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.