The Kranzle Dirt Killer rotary nozzle is engineered to significantly enhance pressure washer effectiveness, providing robust cleaning for diverse hard surfaces. It utilizes a powerful rotating turbine to generate a high-impact whirlwind of water droplets, which delivers five times the cleaning force of a standard flat tip nozzle and efficiently cuts cleaning time in half.
- Rated up to 3200 psi maximum pressure
- Features a 7.0 mm orifice diameter
- Constructed from durable brass, ceramic, and plastic materials
- Includes a 1/4 in NPT female thread for connection
- Withstands inlet water temperatures up to 180 degrees F
- Delivers five times the cleaning impact of a standard flat tip nozzle
- Prevents lines or streaking on cleaned surfaces
- Ideal for blasting away embedded dirt, grime, mold, and mildew