Learn More
PFEIFFER VACUUM INC Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 12, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 6.9 CFM
Catalog No. NC3978937
Encompass_Preferred
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 12, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 6.9 CFM, Noise Level: 50 dB(A), Weight: 27.5kg, Dimensions (L x W x H): 428 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF
Catalog No. NC3978937 Supplier PFEIFFER VACUUM INC Supplier No. 2000239243
Rotary Vane Vacuum Pump, DuoVane, Pfeiffer Vacuum Inc., DuoVane 12, Ultimate Vacuum: 3E-3 mbar, Pumping Speed: 6.9 CFM, Noise Level: 50 dB(A), Weight: 27.5kg, Dimensions (L x W x H): 428 x 164 x 237 mm, Gas inlet/outlet: DN 25 ISO-KF/DN25 ISO-KF