Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Two-Stage Rotary Vane Vacuum Pump, DuoVane 18, 14 m³/h, 31 kg
Supplier: PFEIFFER VACUUM INC 2000239240
The DuoVane 18 is a two-stage, oil-lubricated rotary vane vacuum pump, offering high pumping speeds and efficient operation. Designed for easy process integration, it achieves vacuum levels as low as 3 · 10¯³ hPa(mbar) and is suitable for various industrial and scientific applications.
- Easy installation and operation
- Built-in safety valve prevents gas backflow and uncontrolled venting
- Compact design
- High performance with best pumping-speed-to-size-ratio
- Reduced energy costs and nearly maintenance-free operation
- Optional magnetic coupling prevents leakage and contamination
- All service-relevant components located on one side for easy access
- Universal motor ensures worldwide compatibility